10X Washing Buffer, pH 7.5 Solution, Teknova
Catalog # CA100216-936
Supplier: Teknova
CAS Number:
Specifications
- Description:10X Washing Buffer
- Size:1 L
- Cat. no.:CA100216-936
- pH:7.5
- Supplier No.:W1718
Specifications
About this item
This washing buffer contains 1 M maleic acid and 1.5 M NaCl with pH 7.5.
- Sterile solution
- Suitable for molecular biology applications