Order Entry
Canada
Orders LinkContactUsLinkComponent
Low-Halide Polyester Labels, B30, Brady

Low-Halide Polyester Labels, B30, Brady

Supplier:   Brady Worldwide
Low-Halide Polyester Labels, B30, Brady
Supplier:  Brady Worldwide
Certificates
Low-Halide Polyester Labels, B30, Brady
Low-Halide Polyester Labels, B30, Brady

31 Options Available Below

Product Details & Documents

Hi-Performance Polyester Labels (B-569) have a low halide content and are designed for marking stainless steel pipes, particularly in harsh environments.

  • Durable, ultra-smooth polyester labels with permanent pressure sensitive acrylic adhesive
  • Outdoor grade polyester label can be thermal transfer printed
  • Withstands mild chemicals, oil, and water
  • Service temperature of –40 to 100 °C (–40 to 212 °F)
  • For use with BBP®30; BBP®31; BBP®33; BBP®35; BBP®37 printers

Applies best to very smooth surfaces and ideal for mounting to rigid panels. Continuous roll is not cut to length. It is cut by the printer.

Product Family Options

Product Information

  • Material
    Color
    Width
    Roll length
    Availability
    Price
  • 10018-052
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Brown
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117940
  • 10018-054
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Brown
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117939
  • 10018-056
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Brown
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117938
  • 10018-058
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Brown
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117937
  • 10018-060
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Grey
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117952
  • 10018-450
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Grey
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117951
  • 10018-452
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Grey
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117950
  • 10018-454
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Grey
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117949
  • 10018-456
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Light blue
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117956
  • 10018-458
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Light blue
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117955
  • 10018-460
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Light blue
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117954
  • 10018-462
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Light blue
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117953
  • 10018-464
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Ochre
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117960
  • 10018-466
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Ochre
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117959
  • 10017-994
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Ochre
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117958
  • 10017-996
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Ochre
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117957
  • 10017-998
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Pink
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117968
  • 10018-000
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Pink
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117967
  • 10018-468
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Pink
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117966
  • 10018-470
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Pink
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117965
  • 10018-472
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Purple
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117972
  • 10018-474
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Purple
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117971
  • 10018-476
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Purple
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117970
  • 10018-478
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Purple
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117969
  • 10018-480
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Sky blue
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117980
  • 10018-482
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Sky blue
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117979
  • 10018-484
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Sky blue
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117978
  • 10018-488
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Tan
    0.5"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $271.91
    $271.91
    New Customer Promotional Price
    Each
       $271.91

    Supplier Number: 117984
  • 10018-490
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Tan
    1.125"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $408.34
    $408.34
    New Customer Promotional Price
    Each
       $408.34

    Supplier Number: 117983
  • 10018-492
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Tan
    2.25"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $569.01
    $569.01
    New Customer Promotional Price
    Each
       $569.01

    Supplier Number: 117982
  • 10018-494
    Low-halide polyester labels, B-569
    Low-halide polyester labels, B-569
    Polyester
    Tan
    4"
    30.48 m (100 ft.)
    Log in for availability
    Log in for availability
    Each
    Each - $764.96
    $764.96
    New Customer Promotional Price
    Each
       $764.96

    Supplier Number: 117981
  • Discontinued
    Supplier Number: -
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.