If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Thermal Microbeads, 1 L, 2.5 - 3 mm
- Cat. no.:103302-222
- Supplier No.:68605-10
Specifications
About this item
Effectively heat and cool samples in most water and dry baths.
- Reduce sample contamination in applications requiring water and ice
- Eliminates the need for floating racks and bottle weights
- Autoclavable and solvent resistant
- For use in water/drybaths, ice pans/ buckets, microbead chambers and microbead bag
Use of the microbeads reduces sample contamination in applications requiring water and ice and provides support for tubes and bottles.