Order Entry
Canada
Orders LinkContactUsLinkComponent
Buffer, Sodium Acetate Buffer Solution pH 5,2 (3 mol/L), ROCK

Buffer, Sodium Acetate Buffer Solution pH 5,2 (3 mol/L), ROCK

Sodium Acetate, 3.0M, DEPC Treated, 100 mL, 5.2
Catalog # CAMB-016
Supplier #:  MB-016
Buffer, Sodium Acetate Buffer Solution pH 5,2 (3 mol/L), ROCK
Catalog # CAMB-016
CAS Number:  
Buffer, Sodium Acetate Buffer Solution pH 5,2 (3 mol/L), ROCK

Sodium Acetate, 3.0M, DEPC Treated, 100 mL, 5.2

CAS Number:  
Price
$186.99
Log in for availability
Each (100ml)
Minimum order quantity is 1
Product Details & Documents

 : This product is for research use only and is not intended for therapeutic or diagnostic applications.

Documents

Specifications

Specifications

  • Description:
    Sodium Acetate, 3.0M, DEPC Treated
  • Size:
    100 mL
  • pH:
    5.2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.