Order Entry
Canada
ContactUsLinkComponent
BioPure Silver Nanospheres, 1 mg/ml, Citrate Surface, nanoComposix
BioPure Silver Nanospheres, 1 mg/ml, Citrate Surface, nanoComposix
Catalog # 77679-712
Supplier:  NANOCOMPOSIX, INC.
New Product
undefined
BioPure Silver Nanospheres, 1 mg/ml, Citrate Surface, nanoComposix
Catalog # 77679-712
Supplier:  NANOCOMPOSIX, INC.
Supplier Number:  AGCB5-1M
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Ø:
    5 ±2 nm
  • Surface:
    Citrate (Bare/displaceable surface)
  • Variance:
    <25%
  • Volume:
    1 ml
  • Description:
    BioPure silver nanospheres, 1 mg/ml, in aqueous 2 mM sodium citrate
  • Cat. no.:
    77679-712

Specifications

About this item

Purified, concentrated, high quality silver nanoparticles. Sterile and endotoxin-free. Bare citrate surface.

  • High quality nanoparticles, precisely engineered and highly characterized
  • Unagglomerated and monodisperse
  • High concentration materials (1 mg/mL, 9.27 mM, 1000 ppm, ~40 to 135 OD [depending on size])
  • Extensively purified to remove residual reactants
  • Manufactured sterile; provided in ultra-pure (Milli-Q) DI water
  • BioPure materials ideal for in-vitro and in-vivo experiments, nanotoxicology
  • "Bare" citrate surface is stable and displaceable

Spherical silver nanoparticles, 1 mg/mL (high concentration)

Each batch is extensively characterized to meet highest quality standards, ensuring reliable and reproducible results. All products supplied with COA's including batch-specific TEM, DLS, Zeta, UV-Vis measurements, etc.

Colloidal silver has applications in Diagnostics, Antimicrobial coatings, Electronics and Photovoltaics, SERS, Optics, Nanotoxicology, Plasmonics (SPR/Surface Plasmon Resonance), novel Color Enhancements and Pigments, Molecular Detection, Sensors, and Photothermal and Microwave-based Therapies.

Custom formulations and ISO13485/cGMP compliant materials can be provided on request.

Delivery: Ships via FedEx Priority Overnight (domestic). Cold packaging included. Due to temperature-sensitive nature of these materials, they will only be shipped during the week in order to prevent products from sitting in an uncontrolled temperature environment over the weekend.
Caution: Do not freeze. Store at 2 to 8 °C and away from light when not in use to maximize shelf life. Nanomaterials can be sensitive to high and low pH conditions and high salt concentrations. Use caution when changing environmental conditions as particles may aggregate.