Precision Measurements, RTD Type PTFE-Coated Thermocouple Sensor, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Pt/100 type thermocouple sensors for use with RTD type circulators with LEMO type plugs.
- Stainless steel probes are PTFE-coated
- Temperature range: -200 to 400°C
- Sensors include a 10' lead
Frequently Bought Together