TES buffer, 1X, ready to use, pH 7.8
Catalog # CAMB-058
Supplier: Rockland Immunochemical
CAS Number:
Specifications
- Description:TES, DEPC Treated
- Size:1 L
- Cat. no.:CAMB-058
- pH:7.8
- Supplier No.:MB-058
Specifications
About this item
This product is ready-to-use as a working 1X solution and requires no further dilution
This TES buffer consists of 10 mM Tris HCl, 100 mM NaCl, and 1.0 mM EDTA at a pH of 7.8. Meticulously prepared using ultra pure reagents dissolved in DEPC treated pharmaceutical grade deionized water.
This product was aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.