Annexin V-FITC Apoptosis Detection Kit, Genetex
Catalog # 89280-968
Supplier: Genetex
CAS Number:
Specifications
- Description:Annexin V-FITC Apoptosis Detection Kit
- Size:100 Assay
- Storage conditions:4 °C
- Species reactivity:Mammal
- Cat. no.:89280-968
- Application:FACS, ICC/IF
- Supplier No.:GTX85591
Specifications
About this item
Annexin V-FITC is used for the detection of apoptotic cells (early/middle stages of apoptosis)
- Sample type- Living cells (suspension and adherant)
- Simple one step staining procedure in 10 minutes
- Fast and convenient
- Kit can differentiate apoptosis vs necrosis when performing both Annexin V-FITC and PI staining
For In-vitro laboratory use only. Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.
It also known as Annexin V, Phosphatidylserine Assay, PS Assay, Membrane Apoptosis Assay.
Delivery: It includes Annexin V-FITC, IX Binding Buffer and Propidium Iodide (PI).