Orders LinkContactUsLinkComponent
Radioactive Needle Sources
Radioactive Needle Sources
Radioactive Needle Sources
Certificates
Radioactive Needle Sources
Radioactive Needle Sources
3 Options Available Below
Product Details & Documents
Quick and easy way to show emission with cloud chambers.
  • Safe to use
  • Easy disposal
Needle sources are used to generate a point source of radiation inside cloud chambers for demonstrating alpha and beta radiation tracks. Three different types of isotopes are offered, a pure alpha emitter, a pure beta emitter and a combined alpha/beta emitter.
The sources are constructed by depositing a small, license exempt quantity of radioactive isotope onto the eye of a standard sewing needle which is mounted on a test tube stopper for insertion into the cloud chamber. The needle and stopper are placed into a test tube for protection during shipping and storage.
Product Family Options
Product Information
  • Activity
    Emission
    Half-life
    Availability
    Price
  • 470220-476
    Lead-210 needle source, 0.01 uCi
    Lead-210 needle source, 0.01 uCi
    0.01 μCi
    Alpha (α) Beta (β)
    22.3 years
    Log in for availability
    Log in for availability
    Each
    Each - $71.80
    $71.80
    Each
       $71.80
    Supplier Number: NSPB210
  • 470337-080
    Polonium-210 needle source, 0.01 uCi
    Polonium-210 needle source, 0.01 uCi
    0.01 μCi
    Alpha (α)
    138 days
    Discontinued
    Supplier Number: -
  • 470220-472
    Polonium-210 needle source, 0.01 uCi
    Polonium-210 needle source, 0.01 uCi
    0.01 μCi
    Alpha (α)
    138 days
    Log in for availability
    Log in for availability
    Each
    Each - $71.80
    $71.80
    Each
       $71.80
    Supplier Number: NSPO210
  • 470337-079
    Lead-210 needle source, 0.01 uCi
    Lead-210 needle source, 0.01 uCi
    0.01 μCi
    Alpha (α) Beta (β)
    22.3 years
    Discontinued
    Supplier Number: -
  • 470220-474
    Strontium-90 needle source, 0.01 uCi
    Strontium-90 needle source, 0.01 uCi
    0.01 μCi
    Beta (β)
    28.8 years
    Log in for availability
    Log in for availability
    Each
    Each - $71.80
    $71.80
    Each
       $71.80
    Supplier Number: NSSR90
  • 470337-081
    Strontium-90 needle source, 0.01 uCi
    Strontium-90 needle source, 0.01 uCi
    0.01 μCi
    Beta (β)
    28.8 years
    Discontinued
    Supplier Number: -