Complete Scintillation Probe Assembly
Complete Scintillation Probe Assembly
470220-454
: SPA38
Gamma Spectroscopy detector.
- Unit provides a complete detector assembly for gamma spectroscopy applications
The 38x38 mm NaI(Tl) crystal includes PMT and voltage divider as an integral assembly. The probe is supplied complete with a 10-position sample stand, lead shield and cables for connecting directly to the spectrometer. Resolution is typically 7% @ 662 keV.
: Includes scintillation detector, 38x38mm with voltage divider; GMS38 position stand with sample tray and LS038 lead shield; and BNC6 and MHV6 cable assemblies.
- Description:Complete Scintillation Probe Assembly