Receiver, Moisture Test, Barrett-Type, Chemglass
Supplier: Chemglass
Certificates
About this item
This moisture test receiver is used to determine the water content in petroleum and bituminous material.
- Available with 2mm glass or PTFE stopcock
Receiver is made according to ASTM specifications, with the addition of the top outer standard taper joint and lower stopcock. The 2mL has sub-divisions of 0.02mL; the 5mL has subdivisions of 0.20mL; 10mL has subdivisions of 0.1mL; the 20mL has subdivisions of 0.2 from 0 to 3mL and 0.5 from 3 to 20mL.
Frequently Bought Together