About this item
Designed for your environment, new Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other research and medical products.
- Automatic defrost
- Automatic locking
- Remote alarm contacts
- Quiet operation
- Reduced energy consumption
- Smart View Compatible
Powered by V-drive, the TSX Series delivers low energy, decreased noise levels and improved performance and user experience
Designed to maintain temperature uniformity to protect medical, pharmaceutical-grade products, laboratory media, reagents and more. Unique V-drive technology is designed to save energy without compromising the protection of stored products. U.S. Environmental Protection Agency SNAP (Significant New Alternatives Policy) compliant refrigerants. Quiet operation at just 35dbA (43 during pull down). Sleek exterior design and touch screen. Bright, easy-to-adjust interior LED lighting. Touchscreen controller with password protection. Datalogging with USB download functionality. Battery backup. Choice of reversible glass or solid door. Air or glycol temperature displayport. Audible and visual alarms: power fail, high temp, low temp, low battery, memory full, door ajar. Compatible with most major electronic locks.
These new undercounter refrigerators are quiet and feature an improved user interface, automated locking and increased storage capacity and reliability.
Specifications
- Catalog No:
- 75993-424
- 75993-426
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 75993-424
- 75993-426
- Display:
- Full color TFT-LCD; 4.3"; 480×272
- Full color TFT-LCD; 4.3"; 480×272
- Shelves/drawers:
- 3 full size shelves
- 3 full size shelves
- Refrigerant:
- R744
- R744
- Sound level:
- 35 dBa
- 35 dBa
- Electrical:
- 100 - 240 V, 50 - 60 Hz, 10 A Max./3 A Steady State
- 100 - 240 V, 50 - 60 Hz, 10 A Max./3 A Steady State
- Environmental certification:
- Energy Star; EPA SNAP Compliant
- Energy Star; EPA SNAP Compliant