Polyethylene Bags, Crown Packaging
Catalog #: 11215-131
Supplier: Crown Packaging
Specifications
- W×L:533×610 mm
- Thickness:0.08 mm (0.003")
- Cat. no.:11215-131
- Supplier No.:19047
Specifications
About this item
Polyethylene material.
Customers Who Bought This Also Bought