To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Hard-Sided Field Case
- Cat. no.:89206-424
- Supplier No.:STARA-CS
Specifications
About this item
This hard-sided field case can be used to protect, transport, and store measuring equipment in the field.
The field case holds Star™ A series portable meters (with or without armor), portable electrode holders, extra batteries, solutions, pH electrode in storage sleeve, conductivity probe, and dissolved oxygen probe in storage sleeve. The electrodes and probes can remain connected to meter while in the case, and the internal separating door can be removed for cable management.
Designed for use with Orion™ Star™ A series portable meters (Star A320, Star A220 and Star A120 series).