ContactUsLinkComponent
Orion™ Shorting Strap for BNC Meters, Thermo Scientific
Orion™ Shorting Strap for BNC Meters, Thermo Scientific
Catalog # 34104-823
Orion™ Shorting Strap for BNC Meters, Thermo Scientific
Catalog # 34104-823
Supplier Number:  090045
Specifications
  • Description:
    Orion™ BNC meter shorting strap
  • Cat. no.:
    34104-823
  • Supplier No.:
    090045
Specifications
About this item
Shorting strap for use on pH meters with BNC input to perform BNC input verification testing and meter self test.
The strap may be used with pH meters with BNC input (Orion™ Versa Star™ pH, pH/ISE, and pH/LogR meters; Orion™ Star A series pH, pH/ISE, pH/conductivity, pH/RDO/DO, and pH/ISE/conductivity/RDO/DO meters; Orion™ Dual Star pH/ISE meters; Orion™ 2-Star, 3-Star, 4-Star and 5-Star pH meters).