Order Entry
Canada
Orders LinkContactUsLinkComponent
Sodium hydroxide ≥95.0%, pellets NF

Sodium hydroxide ≥95.0%, pellets NF

Corrosive
Irritant
Sodium hydroxide ≥95.0%, pellets NF
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. 

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • UN:1823
  • ADR:8,II

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay (Total Alkali Including A:
    95.0 - 100.5 %
  • Insoluble Substances & Organic Matter:
    To Pass Test
  • Potassium (K):
    To Pass Test
  • Heavy Metals:
    max. 30 ppm
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $350.17
    $350.17
    Each (500g)
       $350.17

    Supplier Number: SO170-500GM
  • Poly bottle
    6×500 g
    Log in for availability
    Log in for availability
    Case of 6 (1/Itm/Bx)
    Case of 6 (1/Itm/Bx) - $1,156.00
    $1,156.00
    Case of 6 (1/Itm/Bx)
       $1,156.00

    Supplier Number: SO170-500GM-CS
  • Poly bottle
    4×2.5 kg
    Log in for availability
    Log in for availability
    Case of 4 (2,500,g)
    Case of 4 (2,500,g) - $2,319.00
    $2,319.00
    Case of 4 (2,500,g)
       $2,319.00

    Supplier Number: SO170-2.5KG-CS
  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    Each (12kg)
    Each (12kg) - $1,319.68
    $1,319.68
    Each (12kg)
       $1,319.68

    Supplier Number: SO170-12KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.