Order Entry
Canada
ContactUsLinkComponent
Nicotinamide ≥98.5% FCC
Nicotinamide ≥98.5% FCC
Catalog # 75811-750
Supplier:  Spectrum Chemicals
CAS Number:  
Nicotinamide ≥98.5% FCC
Catalog # 75811-750
Supplier:  Spectrum Chemicals
Supplier Number:  N1082-5KG
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly pail
  • Size:
    5 kg
  • Environmentally Preferable:
  • Cat. no.:
    75811-750
  • Supplier No.:
    N1082-5KG

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.5 - 101.0 %
  • Melting Range:
    128 - 131 °C
  • Lead (Pb):
    max. 2 mg/kg
  • Loss On Drying:
    max. 0.5 %
  • Readily Carbonizable Substances:
    To Pass Test

Specification Test Results

About this item

Synonyms: 3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
 :
 :122.13 g/mol
Storage Temperature:Ambient
CAS Number:98-92-0
Formula:C₆H₆N₂O
MDL Number:MFCD00006395