

Hydrochloric acid 37% NF
Supplier: Spectrum Chemical Mfg. Corp.
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.200
Storage Temperature:Ambient
UN:1789
ADR:8,II
:36.46
:GHS05
:GHS07
Specification Test Results
- Assay (w/w):36.5 - 38.0 %
- Bromide or Iodide:To Pass Test
- Free Bromine or Chlorine:To Pass Test
Specification Test Results
Related Information
Certificates
Search by Lot NumberHY104 spec
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together