Order Entry
Canada
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid ≥99.0% USP

L(+)-Ascorbic acid ≥99.0% USP

L(+)-Ascorbic acid ≥99.0% USP
Certificates
L(+)-Ascorbic acid ≥99.0% USP

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Storage Temperature:Ambient

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay:
    99.0 - 100.5 %
  • Specific Rotation [A]D:
    +20.5 to +21.5
  • Residue On Ignition:
    max. 0.1 %
  • Heavy Metals (As Pb):
    max. 20 ppm
  • Identification:
    To Pass Test
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g/mol)
    Each (100g/mol) - $244.52
    $244.52
    Each (100g/mol)
       $244.52

    Supplier Number: AS102-100GM
  • Poly pail
    5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $102,637.19
    $102,637.19
    Each (5kg)
       $102,637.19

    Supplier Number: AS102-5KG
  • Fiber drum
    50 kg
    Log in for availability
    Log in for availability
    Each (50kg)
    Each (50kg) - $3,962.49
    $3,962.49
    Each (50kg)
       $3,962.49

    Supplier Number: AS102-50KGBL

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.