About this item
IntelliCAL™ PHC108 is a robust digital puncture pH electrode with stainless steel body, Ag/AgCl reference system and built-in temperature sensor.
- Compatible with digital Hach meters only (e.g. HQd meters)
- Digital electrode stores time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
- Can be moved between meters without the need to re-calibrate or re-enter measurement settings
- Ideal for a multi-user environment with multiple HQD series laboratory and portable meters
- Ultimate traceability in measurement history
- Non refillable
Ideal for measuring in semi solid samples such as sludge, meats and cheeses.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA10146-090
- Junction:
- Ceramic Pin + Open
- Temperature range:
- 0...60 °C
- Range:
- 2 - 12 pH
- Sensor type:
- Standard