Burette Stands, for TIM8xx, Radiometer Analytical, Hach
Supplier: Hach
Certificates
About this item
High resolution burette stand for TIM8xx series titrators and ION570 benchtop meter.
These stands are UV protected with connection tubing.
Customers Who Bought This Also Bought