Volatile Acid Standard Solution, 62500 mg/L as Acetic Acid, 10 ml, Hach
Catalog # CA1427010
Supplier: Hach
CAS Number:
Specifications
- Concentration:62500 mg/L
- Pack type:16/pack
- Size:10 ml voluette« ampule
- Cat. no.:CA1427010
- Supplier No.:1427010
- Parameters:Volatile Acids as HOAC
Specifications
About this item
For accuracy checks in volatile acids determinations.
- Simple accuracy checks
- Minimize waste
- Reliable results