Order Entry
Canada
ContactUsLinkComponent
Barium Standard Solution, 1000 mg/L as Ba (NIST), 100 mL, Hach
Barium Standard Solution, 1000 mg/L as Ba (NIST), 100 mL, Hach
Catalog # CA71002-746
Supplier:  Hach
CAS Number:  
Barium Standard Solution, 1000 mg/L as Ba (NIST), 100 mL, Hach
Catalog # CA71002-746
Supplier:  Hach
Supplier Number:  1461142
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Size:
    100 ml
  • Cat. no.:
    CA71002-746
  • Certification:
    NIST
  • Element:
    Barium
  • Supplier No.:
    1461142

Specifications

About this item

For accuracy checks in barium determinations.