Order Entry
Canada
ContactUsLinkComponent
Sulfate Standard Solution, 2500 mg/L as SO₄ (NIST), 500 ml, Hach
Sulfate Standard Solution, 2500 mg/L as SO₄ (NIST), 500 ml, Hach
Catalog # CA11028-044
Supplier:  Hach
CAS Number:  
Sulfate Standard Solution, 2500 mg/L as SO₄ (NIST), 500 ml, Hach
Catalog # CA11028-044
Supplier:  Hach
Supplier Number:  1425249
CAS Number:  

Specifications

  • Concentration:
    2500 mg/L SO₄
  • Size:
    500 ml
  • Cat. no.:
    CA11028-044
  • Supplier No.:
    1425249
  • Parameters:
    Sulfate as SO42-(NIST)

Specifications

About this item

For accuracy checks in sulfate determinations.