Cork Borer Sharpener, Humboldt
Catalog # 52924-950
Supplier: Humboldt Manufacturing
Specifications
- Description:Cork borer sharpener
- Cat. no.:52924-950
- Supplier No.:H-9680
Specifications
About this item
The handy tool is used to sharpen borers from 3/16" to 15/16" (5 mm to 24 mm)
The solid brass cone has a hinged steel blade in a slot, and is mounted in a hardwood handle.