Copper Standard Solution, 100 mg/L as Cu (NIST), 100 ml, Hach
Catalog # CA11016-922
Supplier: Hach
CAS Number:
Specifications
- Concentration:100 mg/L Cu
- Size:100 ml
- Cat. no.:CA11016-922
- Supplier No.:12842
- Parameters:Copper as Cu (NIST)
Specifications
About this item
For accuracy checks in copper determinations.