Order Entry
Canada
ContactUsLinkComponent
Silver Nitrate Standard Solution, 0.141 N, 500 ml, Hach
Silver Nitrate Standard Solution, 0.141 N, 500 ml, Hach
  CA11016-518
 :  Hach
 :  
Silver Nitrate Standard Solution, 0.141 N, 500 ml, Hach
  CA11016-518
 :  Hach
 :  1255149
 :  

 

  • Concentration:
    0.141 N
  • Size:
    500 ml
  • Cat. no.:
    CA11016-518
  • Range:
    500 - 25000 mg/L Cl⁻
  • Supplier No.:
    1255149

 

 

For chloride determination by argentometric buret titration.