ContactUsLinkComponent
Ward's® Essentials Exothermic Reactions Lab Kit
  470225-516
undefined
Ward's® Essentials Exothermic Reactions Lab Kit
  470225-516
 :  AD-WE2501-KIT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Exothermic Reactions Lab Kit
  • No. of Lab Groups:
    15
  • Cat. no.:
    470225-516
  • Supplier No.:
    AD-WE2501-KIT
Experience an exothermic reaction and create a heat pack.
  • Hands on lab activity
  • Kit aligned to next generation standards MS-PS1-2
  • Instructions and materials included for 15 groups
  • Middle school
In this lab, students will monitor and graph temperature change in an exothermic reaction, and create a heat pack, which utilizes and exothermic reaction.