Ward's® Essentials Catapult Kit
Catalog # 470225-378
Supplier: Wards
Specifications
- Description:Catapult Kit
- No. of Lab Groups:1
- Cat. no.:470225-378
- Supplier No.:VWRGP00007
Specifications
About this item
Play with the physics of a catapult to learn about levers, force, acceleration and much more!
- Hands on lab activity
- Kit aligned to Next Generation Science Standards MS-PS2-2
- Instructions and materials included for 1 group
- Middle school
Explore force, acceleration, Hooke's law, and parabolic trajectories by building and testing a fully-functional catapult. Contains catapult base, sides, lever arm, rubber bands, screws, dowels, hook, and ping-pong ball.