ContactUsLinkComponent
Pipetting Reservoir, Watson Bio Lab
Pipetting Reservoir, Watson Bio Lab
Catalog # 75982-996
Supplier:  Watson Bio Lab
undefined
Pipetting Reservoir, Watson Bio Lab
Catalog # 75982-996
Supplier:  Watson Bio Lab
Supplier Number:  1791-050-S
Specifications
  • Sterility:
    Sterile
  • Capacity:
    50 ml
  • Description:
    Pipetting Reservoir with Drop-On Cover
  • Cat. no.:
    75982-996
Specifications
About this item
This pipetting reservoir features the Liquid Sweeper.
  • Polypropylene Construction
  • Autoclavable
  • Convenient Graduation
  • Easy-to-Wash Round Covers
  • Liquid Sweeper
Eight grooves and a slope angle make up the Liquid Sweeper, a unique feature that transfers a liquid drop from the top of the tip and gathers liquid without wasting it. The autoclavable polypropylene reservoir also features easy-to-wash round covers and convenient graduation.