Order Entry
Canada
ContactUsLinkComponent
Phosphate Standard Solution, 100 mg/L as PO₄ (NIST), 100 ml, Hach
Phosphate Standard Solution, 100 mg/L as PO₄ (NIST), 100 ml, Hach
Catalog # CA11016-612
Supplier:  Hach
CAS Number:  
Phosphate Standard Solution, 100 mg/L as PO₄ (NIST), 100 ml, Hach
Catalog # CA11016-612
Supplier:  Hach
Supplier Number:  1436832
CAS Number:  

Specifications

  • Concentration:
    100 mg/L PO₄
  • Size:
    100 ml
  • Cat. no.:
    CA11016-612
  • Supplier No.:
    1436832
  • Parameters:
    Phosphate as PO43- (NIST)

Specifications

About this item

For accuracy checks in phosphate (phosphorus) determinations.