Phosphate Standard Solution, 50 mg/L as PO₄ (NIST), 500 ml, Hach
CA17149
: Hach
:
- Concentration:50 mg/L PO₄
- Size:500 ml
- Cat. no.:CA17149
- Supplier No.:17149
- Parameters:Phosphate as PO43- (NIST)
For accuracy checks in phosphate (phosphorus) determinations.