Order Entry
Canada
ContactUsLinkComponent
Iron Standard Solution, 25 mg/L as Fe (NIST), 10 ml, Hach
Iron Standard Solution, 25 mg/L as Fe (NIST), 10 ml, Hach
Catalog # CA11016-760
Supplier:  Hach
CAS Number:  
Iron Standard Solution, 25 mg/L as Fe (NIST), 10 ml, Hach
Catalog # CA11016-760
Supplier:  Hach
Supplier Number:  1425310
CAS Number:  

Specifications

  • Concentration:
    25 mg/L Fe
  • Size:
    10 ml
  • Cat. no.:
    CA11016-760
  • Supplier No.:
    1425310
  • Parameters:
    Iron as Fe (NIST)

Specifications

About this item

For accuracy checks in iron determinations.