ContactUsLinkComponent
Manganese Standard Solution, 1000 mg/L as Mn (NIST), 100 ml, Hach
Manganese Standard Solution, 1000 mg/L as Mn (NIST), 100 ml, Hach
  CA71002-752
 :  Hach
 :  
undefined
Manganese Standard Solution, 1000 mg/L as Mn (NIST), 100 ml, Hach
  CA71002-752
 :  Hach
 :  1279142
 :  
  • Concentration:
    1000 mg/L Mn
  • Size:
    100 ml
  • Cat. no.:
    CA71002-752
  • Supplier No.:
    1279142
  • Parameters:
    Manganese as Mn (NIST)
Standard solutions for accuracy checks in manganese determinations.