Order Entry
Canada
ContactUsLinkComponent
Proteinase K
Proteinase K
Catalog # CA80509-758
Supplier:  MilliporeSigma
CAS Number:  
Proteinase K
Catalog # CA80509-758
Supplier:  MilliporeSigma
Supplier Number:  71049-4
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 ml
  • Storage conditions:
    −20 °C
  • Enzyme name:
    Proteinase K
  • Cat. no.:
    CA80509-758
  • Supplier No.:
    71049-4

Specifications

About this item

Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album.

The enzyme exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA. Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50 to 60 °C). Working concentration is 50 to 100 µg/ml for protein removal and enzyme inactivation and upto 2 mg/ml for tissue treatment. It is a lyophilized powder.