2-Iminothiolane hydrochloride (Traut's Reagent), Pierce™
Catalog # CAPI26101
Supplier: Thermo Scientific
CAS Number:
Specifications
- Size:500 mg
- Cat. no.:CAPI26101
- Supplier No.:26101
Specifications
About this item
:
:137.625 g/mol
Storage Temperature:Refrigerator
CAS Number:4781-83-3
Formula:C₄H₈ClNS
MDL Number:MFCD00039013