Orders LinkContactUsLinkComponent
Disintegration Testing, Quality Lab Accessories
Supplier:   QLA
Disintegration Testing, Quality Lab Accessories
Supplier:  QLA
Certificates
Disintegration Testing, Quality Lab Accessories
Disintegration Testing, Quality Lab Accessories
12 Options Available Below
Product Details & Documents
QLA manufactures Disintegration Testing Accessories and Glassware for Agilent/VanKel disintegration testers.
  • Fluted disks can be used with any brand of disintegration tester
All accessories are USP compliant and made in the USA.
Product Family Options
Product Information
  • Availability
    Price
  • 75848-886
    Disintegration Basket Assembly with 3 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Disintegration Basket Assembly with 3 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Log in for availability
    Log in for availability
    Each
    Each - $2,042.96
    $2,042.96
    Each
       $2,042.96
    Supplier Number: DISASY-03
  • 11010-218
    Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Disintegration Basket Assembly with 6 Glass Tubes and 10 Mesh SS Screens for VanKel Testers
    Log in for availability
    Log in for availability
    Each
    Each - $1,339.37
    $1,339.37
    Each
       $1,339.37
    Supplier Number: DISASY-06
  • 11010-220
    Disintegration Basket Assembly with 6 Plastic Tubes and 10 Mesh SS Screens for VanKel Testers
    Disintegration Basket Assembly with 6 Plastic Tubes and 10 Mesh SS Screens for VanKel Testers
    Log in for availability
    Log in for availability
    Each
    Each - $1,669.77
    $1,669.77
    Each
       $1,669.77
    Supplier Number: DISASY-P6
  • 75848-894
    Glass Replacement Tubes for 3 Tube Disintegration Assembly, 38 mm
    Glass Replacement Tubes for 3 Tube Disintegration Assembly, 38 mm
    Log in for availability
    Log in for availability
    Pack of 3
    Pack of 3 - $311.62
    $311.62
    Pack of 3
       $311.62
    Supplier Number: DISTUB-03
  • 11010-222
    Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Glass Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $180.16
    $180.16
    Pack of 6
       $180.16
    Supplier Number: DISTUB-06
  • 11010-224
    Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Plastic Replacement Tubes for 6 Tube Disintegration Assembly, 25 mm
    Log in for availability
    Log in for availability
    Each
    Each - $646.06
    $646.06
    Each
       $646.06
    Supplier Number: DISTUB-P6
  • 75848-888
    USP Fluted Plastic Disks for 3 Tube Disintegration Assembly
    USP Fluted Plastic Disks for 3 Tube Disintegration Assembly
    Log in for availability
    Log in for availability
    Pack of 3
    Pack of 3 - $245.04
    $245.04
    Pack of 3
       $245.04
    Supplier Number: DISDISK-03
  • 11010-248
    USP Fluted Plastic Disks for 6 Tube Disintegration Assembly
    USP Fluted Plastic Disks for 6 Tube Disintegration Assembly
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $223.31
    $223.31
    Pack of 6
       $223.31
    Supplier Number: FDISC-VK
  • 75848-890
    10 Mesh SS Screens for 3 Tube Disintegration Assembly
    10 Mesh SS Screens for 3 Tube Disintegration Assembly
    Log in for availability
    Log in for availability
    Pack of 3
    Pack of 3 - $90.15
    $90.15
    Pack of 3
       $90.15
    Supplier Number: DISSCR03-VK10
  • 75848-892
    10 Mesh SS Screens for 6 Tube Disintegration Assembly
    10 Mesh SS Screens for 6 Tube Disintegration Assembly
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $90.15
    $90.15
    Pack of 6
       $90.15
    Supplier Number: DISSCR-VK10
  • 11010-348
    Clear Glass Disintegration Beaker with Flared Top, 900 mL
    Clear Glass Disintegration Beaker with Flared Top, 900 mL
    Log in for availability
    Log in for availability
    Each
    Each - $214.05
    $214.05
    New Customer Promotional Price
    Each
       $214.05
    Supplier Number: GLA900-DB
  • 11010-360
    Clear Glass Disintegration Beaker with Flat Top, 900 mL
    Clear Glass Disintegration Beaker with Flat Top, 900 mL
    Log in for availability
    Log in for availability
    Each
    Each - $307.04
    $307.04
    Each
       $307.04
    Supplier Number: GLA905-DB