You searched for: Drum/Transfer Pumps
Drum pumps are efficient solutions for emptying barrels that are too heavy to pour by tipping. Available in a variety of diameters, these fluid transferring devices work quickly to empty tanks, IBCs, or drums. Models such as the siphon, rotary, hand, and piston pumps control the removal of any viscosity level fluid. The drum pumps allow personnel to extract liquids by simple mechanics such as squeezing bulbs or pushing down on buttons.
Masterflex® Motorized Drum Pump Systems, Avantor®
Supplier: VWR International
Complete drum pump system specifically packaged for different applications.
Expand 8 Items
Masterflex® Drum Pump Air Powered Motors, Avantor®
Supplier: VWR International
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
Expand 1 Items
Masterflex® FDA Compliant Sanitary Drum Pumps, Avantor®
Supplier: VWR International
Capable of pumping fluids with viscosities up to 1000 cps.
Expand 9 Items
Masterflex® Drum Pump ODP Motors, Avantor®
Supplier: VWR International
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
Expand 1 Items
Masterflex® Batch Control Drum Pump Systems, Avantor®
Supplier: VWR International
Save money by accurately dispensing expensive fluids — to ±0.5%.
Expand 4 Items
Hand-Operated Chemical-Resistant Drum Pumps
Supplier: VWR International
Safely transfer aggressive chemicals.
Expand 4 Items
Masterflex® Complete Drum and Container Pumps, Avantor®
Supplier: VWR International
System includes pump tube and motor—tube material options ensure chemical compatibility.
Expand 12 Items
Bürkle Polypropylene Hand-Operated Drum Pumps
Supplier: VWR International
Quickly transfer liquids from barrels, canisters, or tanks.
Expand 1 Items
Metal Body Hand-Operated Lever Drum Pumps
Supplier: VWR International
Metal-body pumps for use in tight work areas where drums containing flammables are stored.
Expand 3 Items
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Supplier: VWR International
Dispense liquids safely and quickly from most storage containers.
Expand 3 Items
Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Supplier: VWR International
Capable of pumping fluids with viscosities up to 10,000 cps.
Expand 3 Items
Masterflex® L/S® Portable Sampling Pumps, Avantor®
Supplier: Avantor Fluid Handling
Sample depths down to 7.9 meters (26 feet).
Expand 1 Items
Bath Draining Siphon, Grant Instruments
Supplier: GRANT USA INC.
Bath draining siphon for use with Grant Instruments water baths.
Expand 1 Items
Supelco® Solvent Pumps, Merck
Supplier: MilliporeSigma
Supelco® Solvent Pumps, Merck
Expand 3 Items
ELGA Pumps for Water Purification Systems, ELGA LabWater
Supplier: ELGA CA
By choosing an ELGA water purification system, you are making an investment in quality, reliability and value.
Expand 2 Items
PTFE Barrel Pumps, Ultrapure, Burkle
Supplier: BURKLE INC
Due to the high chemical resistance of the medium-contacting parts made of PTFE and FEP, even liquids which attack conventional materials such as PP or PVC can now be safely transferred (except for elemental fluorine) with this barrel pump. The barrel pump is also suitable for transferring ultra-pure liquids. The stopcock prevents hazardous vapors from escaping, and stops dripping after transfer.
Expand 6 Items
Pump-It® Container Pump, Bürkle
Supplier: BURKLE INC
Pump-It® Container Pump, Bürkle
Expand 1 Items
SP Bel-Art Poly-Hand Pumps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
The Poly-Hand pump threads directly to a standard drum bung or carboy.
Expand 3 Items
Nylon Hand Pump, New Pig
Supplier: New Pig
Nylon hand pumps fits 15- through 55-gallon drums with 2" NPT bungs.
Expand 1 Items
Gas-Tight Barrel Pump, Polypropylene, Bürkle
Supplier: BURKLE INC
The Gas-Tight Barrel Pump enables odor-free transfer.
Expand 2 Items
Drum Pumps, Bürkle
Supplier: BURKLE INC
Drum Pumps made of PP available with rigid discharge tube or flexible PVC discharge hose with stopcock
Expand 12 Items
Foot-Operated Solvent Pumps, Bürkle
Supplier: BURKLE INC
Foot-Operated Solvent Pumps, Bürkle
Expand 2 Items
PumpMaster Container Pumps, Burkle
Supplier: BURKLE INC
The manual pressure pump is ideally suited for drawing off, decanting and emptying canisters and barrels.
Expand 3 Items
Drum Pumps for Combustible Liquids, Bürkle
Supplier: BURKLE INC
Drum Pumps for Combustible Liquids made of Stainless steel V2A (1.4301) / AISI 304 available with rigid discharge tube or flexible 1.2 m PTFE/stainless steel discharge hose with ball valve made of stainless steel
Expand 6 Items
Gas-Tight Stainless Steel Drum Pumps, Burkle
Supplier: BURKLE INC
Gas-tight barrel pump with flexible discharge hose for filling off into large canisters and containers.
Expand 1 Items
Dosing Pumps, Dosi-Pump, Bürkle
Supplier: BURKLE INC
Dosi pump are hand controlled pump used for the controlled product extraction from canisters, bottles, barrels and carboys
Expand 5 Items
Oil Safe® Pump, PIG®
Supplier: New Pig
The Oil Safe® Pump dispenses lubricants in hard-to-reach spots.
Expand 1 Items
SP Bel-Art Vacuum Aspirator Pump, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
High-density polyethylene aspirator pump operates on water pressure above 11psi (7584 N/m2).