SilenceMag™ Transfection Reagent, OZ Biosciences
Catalog # 103549-568
Supplier: OZ Biosciences
CAS Number:
Specifications
- Cell specific:Hard-to-transfect
- dsRNA:Yes
- siRNA:Yes
- Description:SilenceMag Super Starting Kit
- Size:Super Magnetic Plate + 100 µL PolyMag + 100 µL CombiMag + 100 µL PolyMag Neo + 200 µL SilenceMag
- Cat. no.:103549-568
- No. of Reactions:400
Specifications
About this item
SilenceMag siRNA Delivery Reagent is based on the Magnetofection technology.
- High protein knockdown efficiency
- For all siRNA applications
- No off-target effects
- Serum compatible and non toxic
- Simple, rapid, and easy-to-use
Simple, rapid and easy to use, SilenceMag is serum compatible and non toxic. Specifically designed for siRNA transfection, SilenceMag gives reliable high protein knockdown at very low doses of siRNA in numerous cell types (primary cells, hard-to-transfect & cell lines).
Accessories information: This reagent needs to be used with a magnetic plate
Delivery: Shipped at room temperature
Packaging: Store at +4 °C