Free Fatty Acid Fluorometric Assay Kit, Cayman Chemical Company
Catalog # 75817-130
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Cat. no.:75817-130
- Includes:Kit contains: Standard, Fluorometric Detector, Assay Buffers, Reagents, and 96-Well Plate
- Supplier No.:700310-96
Specifications
About this item
A sensitive tool for measuring free fatty acids in plasma, serum, and urine.
- Measure FFAs in plasma, serum, and urine
- Assay 20 samples in duplicate
- Measure free fatty acids from 25 to 250 µM
- Plate-based fluorometric measurement (ex 530 - 540 nm, em 585 - 595 nm)
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Fluorometric Assay provides a simple, reproducible, and sensitive tool for measuring free fatty acids in plasma, serum, and urine. The FFA Fluorometric Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.