Order Entry
Canada
ContactUsLinkComponent
Nitrate/Nitrite Colorimetric Assay Kit, Cayman Chemical Company
Nitrate/Nitrite Colorimetric Assay Kit, Cayman Chemical Company
Catalog # 75817-230
CAS Number:  
undefined
Nitrate/Nitrite Colorimetric Assay Kit, Cayman Chemical Company
Catalog # 75817-230
Supplier Number:  780001-192
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Cat. no.:
    75817-230
  • Range:
    5 - 35 µm
  • Includes:
    Kit contains: Standards, Buffer, Reagents, and 96-Well Plates
  • Supplier No.:
    780001-192

Specifications

About this item

For the measurement of total nitrate/nitrite concentrations.

  • Measure nitrate, nitrite, and/or total nitrate/nitrites in plasma/serum, urine, tissue culture media, and tissue homogenates
  • Assay 40 samples in duplicate/96-well plate
  • Assay Range: 5-35 µM
  • Plate-based colorimetric measurement (540-550 nm)

The Cayman Chemical Nitrate/Nitrite Assay Kit provides an accurate and convenient method for measurement of total nitrate/nitrite concentration in a simple two-step process

This kit can be used to measure nitrate and nitrite in plasma/serum, urine, tissue culture media, and tissue homogenates. However, it cannot be used to analyze nitrate and nitrite from an in vitro assay of nitric oxide synthase (NOS) in which excess NADPH has been added. For these assays a second method (LDH method) is utilized and is available from Cayman Chemical in a 96 well plate format.

The first step is the conversion of nitrate to nitrite utilizing nitrate reductase. The second step is the addition of the Griess Reagents which convert nitrite into a deep purple azo compound. Photometric measurement of the absorbance due to this azo chromophore accurately determines nitrite concentration.