HyFlex® 11-270 Tube Knit Nylon Cut-Resistance Sleeve, without Thumb-Slot, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Advanced mechanical protection economical light duty cut resistant sleeve.
- The new HyFlex® 11-270 sleeves are made of double-layered nylon for a soft and comfortable wear experience
- Seamless design and multiple length options ensure an excellent fit and protection
- Made to perform in combination with any HyFlex® cut-resistant glove
HyFlex® 11-270 is the ideal daily sleeve for light mechanical protection
HyFlex® 11-270 sleeves are ideally suited for light assembly, logistics, warehousing, and general handling applications in automotive and transport, aerospace, machinery and equipment, metal fabrication, and white goods industries.
The wrist and elastic band at the top provide stability for the forearms. These ambidextrous sleeves feature a dirt masking colour that enables prolonged wear. They are silicone-free for use in paint and finish processes.
Certifications: ANSI Cut Level 2.
Caution: Not recommended for use with serrated or powered cutting tools, or motorized blades.
Specifications
- Catalog No:
- 75845-976
- 75845-978
- Model:
- HyFlex® 11-270
- HyFlex® 11-270
- Garment application:
- Industrial, Laboratory
- Industrial, Laboratory
- Gender:
- Unisex
- Unisex
- Fabric:
- Nylon
- Nylon