You searched for: Prepared Media for Microbiology
Prepared microbiology media support accurate microorganism detection, identification, and cultivation. Saving time by eliminating the requirement to complete the groundwork, the media increase the overall working efficiency for any laboratory. Minimizing the possibility of formula variability, the standardized solutions ensure that data errors do not occur. The prepared microbiology media are plated or contained in tubes to guarantee that the correct nutrient mixtures are present for specific cell types.
Prepared Nutrient Broth Tubes
Supplier: Culture Media & Supplies
NUTRIENT BROTH PKG/12 TUBES
Expand 2 Items
Yeast Genetic (MIN) Agar
Supplier: IPM SCIENTIFIC, INC.
For Demonstration of Typtophan Dependance or Independence in Yeast
Expand 1 Items
Ward's® Prepared Yeast, Genetic (YEAD) Agar
Supplier: Culture Media & Supplies
Ward's® Prepared Yeast, Genetic (YEAD) Agar
Expand 2 Items
VWR® LB Broth Liquid
Supplier: VWR
LB Broth, Liquid is used for the propagation and maintenance of E.coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
Expand 1 Items
VWR® SOC Outgrowth Media, Biotechnology Grade
Supplier: VWR
SOC Media is a rich media that increases transformation efficiency in competent E. coli. After transformation, cells are incubated in SOC to allow expression of antibiotic resistance genes prior to selection.
Expand 1 Items
Prepared Nutrient Agar
Supplier: Avantor
General-purpose media support the growth of a variety of bacteria and fungi.
Expand 4 Items
MacConkey Agar
Supplier: IPM SCIENTIFIC, INC.
For Differential Isolation of Enteric Bacilli Based on Lactose Fermentation
Expand 1 Items
1.0% Agarose, 200 ml
Supplier: Ward's Science
Jump to the electrophoresis step quicker - let us prepare your gel for you.
Expand 3 Items
Starch Agar
Supplier: IPM SCIENTIFIC, INC.
This media can be used to determine if a microorganism is capable of hydrolyzing starch as a source of energy for growth.
Expand 1 Items
Sordaria Mating Agar
Supplier: Culture Media & Supplies
For the Study of Genetic Crossover
Expand 2 Items
Yeast, Genetic (YED) Agar, Plates
Supplier: IPM SCIENTIFIC, INC.
AGAR (COMPLETE YED) PKG/10 PLATES
Expand 1 Items
Triple Sugar Iron (TSI) Agar Tubes
Supplier: Avantor
Triple Sugar Iron (TSI) Agar Tubes
Expand 1 Items
Prepared Nutrient Agar Deeps, Tubes
Supplier: Avantor
Contains 12 nutrient agar deeps. Used to check for organism motility. 17 mL per tube.
Expand 1 Items
Ward's® Luria Agar Media Plates and Pour Packs
Supplier: Culture Media & Supplies
All-purpose medium that supports the growth of a variety of microorganisms.
Expand 6 Items
Microbiological Media in Ampoules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
A wide variety of sterile ampoule media for common microbiological analyses, in clear glass or plastic ampoules. Supplied pre-measured for maximum efficiency and convenience. For public wastewater and drinking water: MF-Endo and M-FC (with and without rosolic acid for helping to increase the specificity of the medium for faecal coliforms) media are used when analysing water samples for total coliforms, faecal coliforms and E. coli. For food and beverage: M-TGE, HPC, M-Green YM, and orange serum broths are used in the food and beverage industry to QC final products (and fluids used in the manufacture of these products) to detect potential spoilage organisms. Trypticase soy broth, M-TGE, and HPC may be used to test final products and process waters for total bacteria in the pharmaceutical and microelectronics industries. Pseudomonas and KF-Streptococcal broths are highly selective media for organisms in these genera, and are used to monitor samples wherever contamination by these organisms must be prevented.
Expand 13 Items
Actero™ Sabouraud Dextrose Agar with Chloraphenicol
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Expand 12 Items
Actero™ Lactobacilli MRS Broth (LMRS Broth)
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Expand 27 Items
ACTERO™ UVM1 Complete Media
Supplier: Foodchek
The Actero™ MediaBox™ is the solution for laboratories that require liquid culture media on-demand
Expand 27 Items
Actero™ Listeria Enrichment Media
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing enabling targetl pathogen present to grow and be detected in test samples
Expand 23 Items
Actero™ EC Medium
Supplier: Foodchek
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Expand 9 Items
Coliscreen, Hardy Diagnostics
Supplier: Hardy Diagnostics
For the presumptive identification of Escherichia coli.
Expand 1 Items
LB Broth (Lennox), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
LB Broth, Lennox is a ready-to-use microbial growth medium used for the cultivation of E. coli.
Expand 2 Items
LIA (Lysine Iron Agar) Agar
Supplier: Hardy Diagnostics
Lysine Iron Agar are used for the identification of gram negative enteric pathogens.
Expand 1 Items
Viral transport medium
Supplier: Hardy Diagnostics
Hardy Diagnostics Viral Transport Medium is recommended for the collection and transport of clinical specimens for the recovery of viral agents.
Expand 1 Items
Tryptic Soy Agar Plates with 5% Sheep Blood, Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
Trypticase Soy Agar with 5% Sheep Blood is used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species
Expand 1 Items
Corning® PureCoat™ Collagen I, Rectangular Straight Neck Cell Culture Multi-Flask, 3-Layer with Vented Cap, Corning
Supplier: Corning
Synthetic peptide, animal-free, USP class VI polystyrene suitable for cell culture.
Expand 2 Items
Czapek Dox Agar, Deep Fill
Supplier: HARDY DIAGNOSTICS CA
Accommodates deeper fills. Used for the cultivation of saprophytic fungi and soil bacteria capable of utilizing inorganic nitrogen.
Expand 2 Items
Prepared media (BHI agar with blood slant)
Supplier: Hardy Diagnostics
Brain Heart Infusion Agar with 5% Sheep Blood for the cultivation of fungi.