SP Wilmad-LabGlass Barrett Moisture Traps, SP Industries
Supplier: SP Industries
Certificates
About this item
Calibrated “to contain” at 20°C, these distilling receivers are used for determination of water in various materials by distillation with a solvent that is not miscible with water.
- Borosilicate glass
With full length Standard Taper inner member for connection to distilling flask and a Standard Taper outer joint of the same size to connect the condenser, these moisture traps are supplied with a 2mm stopcock.
Customers Who Bought This Also Bought