About this item
These environmentally friendly ovens save energy and lower CO2 emission.
- Microprocessor PID Control
- Two Convenient Exhaust Ports
- Heat-up Time up to 15 Minutes Shorter than Previous Models
- Forced Air Circulation Method
Heat tightness and insulation design of the chamber helps the ovens save 30% of energy during constant temperature compared to previous models
Users can adjust the temperature setting with a digital key. Operation functions include fixed-temperature operation and auto stop and start. Display functions include integration time, calendar time, calibration offset, power consumption, CO2 discharge, heater operation, and time display. Safety features such as self-diagnostic functions, a key lock function, and an electric leakage breaker with over-current protection are provided.
A TRIAC zero-cross control manages the heater circuit. Ovens feature one Scirocco fan, and the largest unit features two. They also feature a stainless pipe heater, and the largest two units have two each. The ovens' interior is made of stainless steel, and the exterior features electro-galvanized steel sheet with epoxy and melamine-resin baking finish. Brackets and a shelf are provided.
Specifications
- Catalog No:
- 75846-534
- 75846-530
- Insulation:
- Glass Wool
- Glass Wool
- Max. load per shelf:
- ~15 kg
- ~15 kg
- Sensor:
- K Thermocouple
- K Thermocouple
- Timer:
- 1 Minute to 99 Hours, 59 Minutes/1 Minute
- 1 Minute to 99 Hours, 59 Minutes/1 Minute