ContactUsLinkComponent
QuantiFluo™ Protein Assay Kit, BioAssay Systems
QuantiFluo™ Protein Assay Kit, BioAssay Systems
Catalog #: 75878-192
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
KIT PRT ASY QUANTIFLUO 50 NG/ML 200 TEST
QuantiFluo™ Protein Assay Kit, BioAssay Systems
Catalog #: 75878-192
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Protein Assay Kit
  • Size:
    200 assays
  • Method:
    FL360/450 nm
  • Sample:
    Biological
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-192
  • Supplier No.:
    QFPR-200
Specifications
About this item
For quantitative fluorimetric determination of total protein.
  • Linear detection range of 0.05 - 200 μg/mL BSA.
  • Homogeneous "mix-incubate-measure" type assay Can be readily automated to test thousands of samples per day
The protein or peptide is known as the "building blocks of life" and is one of the most important macromolecules in life science
BioAssay Systems’ QuantiFluo™ protein assay kit is based on an improved o-phthalaldehyde method. This reagent reacts with primary amines in protein or peptide and forms a blue fluorescent product, allowing detection of nanograms of proteins. The fluorescence intensity (λex/em = 360/450nm) is proportional to the protein concentration in the sample.
Protein determination is a very common practice. Simple, direct and automation-ready. Procedures for measuring protein or peptide concentration are very desirable
Customers Who Bought This Also Bought