ContactUsLinkComponent
QuantiFluo™ HRP Detection Reagent, BioAssay Systems
QuantiFluo™ HRP Detection Reagent, BioAssay Systems
Catalog # 75878-124
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ HRP Detection Reagent, BioAssay Systems
Catalog # 75878-124
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  QFHRP-25ML
CAS Number:  
Specifications
  • Description:
    HRP Detection Reagent
  • Size:
    25 mL
  • Method:
    OD570 nm, FL530/585 nm
  • Sample:
    Biological, compatible with all species
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-124
  • Supplier No.:
    QFHRP-25ML
Specifications
About this item
Stabilized 10-Acetyl-3,7-dihydroxyphenoxazine (also called Ampliflu Red) liquid substrate reagent for HRP detection.
  • For both absorbance or fluorescence detection of horse-radish peroxidase activity
  • Highly sensitive, convenient and fast
QuantiFluo™ Horseradish Peroxidase Reagent is a stable 10-Acetyl-3,7-dihydroxyphenoxazine (also called Ampliflu Red) liquid substrate reagent formulated for detecting horseradish peroxidase HRP activity, e.g. in ELISA. This reagent produces a highly fluorescent product (λex/em = 530/590nm) when reacted with H2O2 by a peroxidase. The fluorescent product can also be visualized under a light source (530 ± 20 nm).