QuantiFluo™ ALP Detection Reagent, BioAssay Systems
Catalog # 75878-190
Supplier: BIOASSAY SYSTEMS
CAS Number:
Specifications
- Description:ALP Detection Reagent
- Size:6 mL
- Method:FL360/450 nm
- Sample:Biological
- Cat. no.:75878-190
- No. of Reactions:600
- Supplier No.:QFALP-6ML
Specifications
About this item
Stabilized 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent for detecting alkaline phosphatase activity.
- Stable single reagent
- Highly sensitive fluorescence detection
- Can be visualized under a UV light
QuantiFluo™ ALP Reagent is a stable 4-methylumbelliferyl phosphate (MUP) liquid substrate reagent formulated for detecting alkaline phosphatase ALP, e.g. in ELISA. This single reagent produces a highly fluorescent product (λex/em = 360/450 nm) when dephosphorylated by ALP. The fluorescent product can also be visualized under a UV light.