Order Entry
Canada
ContactUsLinkComponent
EnzyChrom™ Phospholipase D Assay Kit, BioAssay Systems
EnzyChrom™ Phospholipase D Assay Kit, BioAssay Systems
Catalog # 75878-162
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyChrom™ Phospholipase D Assay Kit, BioAssay Systems
Catalog # 75878-162
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EPPD-100
CAS Number:  

Specifications

  • Description:
    Phospholipase D Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-162
  • Supplier No.:
    EPPD-100

Specifications

About this item

For quantitative determination of phospholipase D activity.

  • Detection range colorimetric assay 0.06 - 10 U/L, fluorimetric assay 0.04 - 1 U/L
  • Can be readily adapted to high-throughput assays for drug screening

Phospholipase D (PLD) catalyses the hydrolysis of the phosphodiester bond of glycerophospholipids to generate phosphatidic acid and a free headgroup

BioAssay Systems’ method provides a simple and high-throughput assay for measuring PLD activity. In this assay, PLD hydrolyzes phosphatidylcholine to choline which is determined using choline oxidase and a H2O2 specific dye. The optical density of the pink colored product at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the PLD activity in the sample.

Abnormalities in PLD expression have been associated with human cancers.