ContactUsLinkComponent
EnzyChrom™ Glutathione Assay Kit, BioAssay Systems
EnzyChrom™ Glutathione Assay Kit, BioAssay Systems
Catalog #: 75878-138
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
KIT GSH/GSSG ASY ENZYCHROM 10 NM 100TEST
EnzyChrom™ Glutathione Assay Kit, BioAssay Systems
Catalog #: 75878-138
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glutathione Assay Kit
  • Size:
    100 assays
  • Method:
    OD412nm
  • Sample:
    Whole blood, plasma, serum, urine, tissue and cell extracts
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-138
  • Supplier No.:
    EGTT-100
Specifications
About this item
For quantitative determination of reduced and oxidized glutathione (GSH/GSSG).
  • Linear detection range 0.01-3 µM GSH equivalents
Glutathione, a tripeptide of glycine, glutamic acid and cysteine, is one of the key antioxidants involved in protecting cells from damages by reactive oxygen species
BioAssay Systems’ GSH/GSSG Assay Kit is designed to accurately measure total, reduced and oxidized glutathione in biological samples using an enzymatic method that utilizes Ellman’s Reagent (DTNB) and glutathione reductase (GR). DTNB reacts with reduced glutathione to form a yellow product. The rate of change in the optical density, measured at 412 nm, is directly proportional to glutathione concentration in the sample. This kit can also be used to measure oxidized (GSSG) by using a specific protocol which first scavenges all GSH with 1-methyl-2-vinylpyridinium triflate.
Glutathione (GSH) reduces disulfide bonds in cytoplasmic proteins to cysteines, in which it is converted to its oxidized form GSSG.
Customers Who Bought This Also Bought